CPC® AseptiQuik® L Series Genderless Barb Aseptic Connectors
Supplier: Colder Products
AQL17012 AQL17012HT AQL17016 AQL17016HT
MFLX20202-58EA 314.42 CAD
MFLX20202-58 MFLX20202-59 MFLX20202-60 MFLX20202-61
CPC® AseptiQuik® L Series Genderless Barb Aseptic Connectors
Fittings and Connectors
Provide easy, secure sterile connections even in nonsterile environments now in newly engineered larger sizes.
  • Genderless design simplifies system integration—no need to worry about female and male threads
  • Robust construction provides reliable performance—even without clamps
  • Quick, three-step connection process reduces risk of operator error
Manufactured from sturdy polycarbonate, these genderless connectors are ideal for single-use and sanitary applications. Use hose barb connectors with filters, tubing, and process containers to create systems forMore Product Information