Masterflex® Single-Use Bag Port Plate Ribbed Base, Avantor®
Supplier: VWR International
MFLX40332-00 MFLX40332-01 MFLX40332-02 MFLX40332-03 MFLX40332-04 MFLX40332-05 MFLX40332-06 MFLX40332-07 MFLX40332-08 MFLX40332-09
MFLX40332-00EA 59.41 CAD
MFLX40332-00 MFLX40332-01 MFLX40332-02 MFLX40332-03 MFLX40332-04 MFLX40332-05 MFLX40332-06 MFLX40332-07 MFLX40332-08 MFLX40332-09
Masterflex® Single-Use Bag Port Plate Ribbed Base, Avantor®
Fittings and Connectors
Ribbed base prevents suction blockage and increases rigidity.
  • Barb sizes range from 1/8" to 1"
  • 1-3/4" and 2-1/4" ribbed bases
Masterflex® Single-Use Bag Ports are engineered to be thermal welded or ultrasonic welded to medical and bioreactor bags of similar material. These materials include polyethylene. These bags can be used to transfer fluid, drug compounds, and cell culture in the biopharmaceudical markets. If material validation is needed, please reach out to our techincal support representatives.