Masterflex® Single-Use Bag Port Disc Flat Plate
Supplier: Avantor Fluid Handling
Masterflex®
PND4-E8402-QC-MF-10PK PND3-E8402-QC-MF-10PK PND45-6-E8402-MF-10PK PND10-E8402-QC-MF-10PK PND2.5-E8402-QC-MF-10PK PND8-E8402-QC-MF-10PK PND6-E8402-QC-MF-10PK PND12-E8402-QC-MF-10PK PND-LF E8402-QC-MF-10PK PND2-E8402-QC-MF-10PK PND5-E8402-QC-MF-10PK PND16-E8402-QC-MF-10PK
MFLX40331-25EA 56.09 CAD
MFLX40331-25 MFLX40331-26 MFLX40331-16 MFLX40331-27 MFLX40331-17 MFLX40331-18 MFLX40331-19 MFLX40331-20 MFLX40331-21 MFLX40331-22 MFLX40331-23 MFLX40331-24
Masterflex® Single-Use Bag Port Disc Flat Plate
Fittings and Connectors
Flexible, smooth flat base.
  • Barb sizes range from 1/8" to 1"
  • 2-1/4" disc diameter
Masterflex® Single-Use Bag Ports are engineered to be thermal welded or ultrasonic welded to medical and bioreactor bags of similar material. These materials include polyethylene. These bags can be used to transfer fluid, drug compounds, and cell culture in the biopharmaceudical markets. If material validation is needed, please reach out to our techincal support representatives.