Masterflex® Barbed Micro2 Flow Open Male Quick-Disconnect Fittings, Avantor®
Supplier: VWR International
MFLX40262-14EA 60.67 CAD
MFLX40262-14 MFLX40262-16 MFLX40262-17 MFLX40262-18 MFLX40262-19 MFLX40262-20 MFLX40262-21
Masterflex® Barbed Micro2 Flow Open Male Quick-Disconnect Fittings, Avantor®
Fittings and Connectors
Mechanical innovations reduce risk of fluid cross-over and contamination.
  • Aseptic connection and disconnection
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
  • A complete coupler requires a male and female fitting with same flow size (order separately). Compatible with Female Micro2 Quick-Disconnect fittings.
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to passMore Product Information