Masterflex® Barbed Micro Flow Male Quick-Disconnect Fittings, Avantor®
Supplier: VWR International
MFLX40261-08 MFLX40261-09 MFLX40261-10 MFLX40261-11 MFLX40261-12 MFLX40261-13 MFLX40261-14 MFLX40261-15 MFLX40261-16 MFLX40261-17 MFLX40261-18 MFLX40261-19 MFLX40261-20 MFLX40261-21 MFLX40261-22 MFLX40261-23
MFLX40261-08EA 29.7 CAD
MFLX40261-08 MFLX40261-09 MFLX40261-10 MFLX40261-11 MFLX40261-12 MFLX40261-13 MFLX40261-14 MFLX40261-15 MFLX40261-16 MFLX40261-17 MFLX40261-18 MFLX40261-19 MFLX40261-20 MFLX40261-21 MFLX40261-22 MFLX40261-23
Masterflex® Barbed Micro Flow Male Quick-Disconnect Fittings, Avantor®
Fittings and Connectors
Mechanical innovations reduce risk of fluid cross-over and contamination.
  • Aseptic connection and disconnection
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
  • A complete coupler requires a male and female fitting with same flow size (order separately). Compatible with Female Micro Quick-Disconnect fittings.
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass throughMore Product Information