Masterflex® Barbed Micro Flow Female Quick-Disconnect Fittings, Avantor®
Supplier: VWR International
MFLX40261-00 MFLX40261-01 MFLX40261-02 MFLX40261-03 MFLX40261-04 MFLX40261-05 MFLX40261-06 MFLX40261-07
MFLX40261-00EA 46.77 CAD
MFLX40261-00 MFLX40261-01 MFLX40261-02 MFLX40261-03 MFLX40261-04 MFLX40261-05 MFLX40261-06 MFLX40261-07
Masterflex® Barbed Micro Flow Female Quick-Disconnect Fittings, Avantor®
Fittings and Connectors
Mechanical innovations reduce risk of fluid cross-over and contamination.
  • Aseptic connection and disconnection
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
  • A complete coupler requires a male and female fitting with same flow size (order separately).
  • Compatible with Male Micro Quick-Disconnect fittings.
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass throughMore Product Information