Masterflex® 1½ Sanitary Tri-Clamp to 1½" Sanitary Tri-Clamp", Avantor®
Supplier: VWR International
SF16-SF16-NK7-QC-MF 10PK SF16-SF16-PP-QC-MF 10PK
MFLX40330-42EA 316 CAD
MFLX40330-42 MFLX40330-43 MFLX40330-44
Masterflex® 1½ Sanitary Tri-Clamp to 1½" Sanitary Tri-Clamp", Avantor®
Fittings and Connectors
Maintain sanitary conditions while easily and securely connecting process lines.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Meet requirements for USP Class VI, FDA, REACH, and California Proposition 65
  • Compatible with other Tri-Clamp® connections
Tri-Clamp® unions allow for secure connection of sanitary lines. Though both materials may be autoclaved, PVDF tolerates high temperatures better than Polypropylene does and will perform more consistently after repeated autoclaving. Clamps and gaskets required for a complete system are sold separately.