Masterflex® Female Luer to Barb Fitting, Avantor®
Supplier: VWR International
LF-1.5CVU-QC-MF-10PK LF-1.5NK7-QC-MF-10PK LF-1.5PP-QC-MF-10PK LF-1MN-QC-MF-10PK LF-1NK7-QC-MF-10PK LF-1NN-QC-MF-10PK LF-1PP-QC-MF-10PK LF-2.5NN-QC-MF-10PK LF-2MN-QC-MF-10PK LF-2PP-QC-MF-10PK LF-3CVU-QC-MF-10PK LF-3NK7-QC-MF-10PK LF-4CVU-QC-MF-10PK LF-4TPE-QC-MF-10PK
MFLX40314-00EA 18.17 CAD
MFLX40314-00 MFLX40314-01 MFLX40314-02 MFLX40314-03 MFLX40314-04 MFLX40314-05 MFLX40314-06 MFLX40314-07 MFLX40314-08 MFLX40314-09 MFLX40314-10 MFLX40314-11 MFLX40314-12 MFLX40314-13 MFLX40314-14 MFLX40314-15 MFLX40314-16 MFLX40314-17 MFLX40314-18 MFLX40314-19 MFLX40314-20 MFLX40314-21 MFLX40314-22 MFLX40314-23 MFLX40314-24 MFLX40314-25 MFLX40314-26 MFLX40314-27 MFLX40314-28 MFLX40314-29 MFLX40314-30
Masterflex® Female Luer to Barb Fitting, Avantor®
Fittings and Connectors
Easily transition to a luer fitting.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Determine which connector type is needed for your application by contacting free technical application assistance
These fittings are manufactured in an ISO Class 7 cleanroom at an ISO 9001 facility; utilizing an ISO 13485 quality system. There is full lot traceability and tight dimensional tolerances for increased reliability and process purity. All female ports may be used with either male slip or male luer lock connections. Utilizing a wide range of fitting materials, we can support applications requiring specific chemical compatibility.
CrystalVu:
Sterilization: Gamma irradiation, Ethylene Oxide, Autoclave
Regulatory Approvals: USP Class VI, FDA 21 CFR 177.1520
Medical Nylon:
Sterilization: Ethylene Oxide, Autoclave
Regulatory Approvals: FDA 21 CFR 178.3297
Natural Nylon:
Sterilization: Ethylene Oxide, Autoclave
Regulatory Approvals: FDA 21 CFR 177.1500
Natural Kynar:
Sterilization: Gamma irradiation, Ethylene Oxide, Autoclave
Regulatory Approvals: USP Class VI, FDA 21 CFR 177.2510
Polypropylene:
Sterilization: Gamma irradiation
Regulatory Approvals: FDA 21 CFR 178.3297, USP Class VI
USP Class VI applies to one of the six classifications for plastics from General Chapter 88 of the United States Pharmacopeia and National Formulary (USP-NF)
Class VI is deemed the most rigorous and thorough