Masterflex® 3/4" Sanitary Clamp High Flow Male Quick-Disconnect Fittings, Avantor®
Supplier: VWR International
MFLX40257-44 MFLX40257-45 MFLX40257-46 MFLX40257-51 MFLX40257-52 MFLX40257-53 MFLX40257-58 MFLX40257-59 MFLX40257-60 MFLX40257-65 MFLX40257-66 MFLX40257-67 MFLX40257-72 MFLX40257-73 MFLX40257-74 MFLX40257-80 MFLX40257-81 MFLX40257-82
MFLX40257-44EA 143.15 CAD
MFLX40257-44 MFLX40257-45 MFLX40257-46 MFLX40257-51 MFLX40257-52 MFLX40257-53 MFLX40257-58 MFLX40257-59 MFLX40257-60 MFLX40257-65 MFLX40257-66 MFLX40257-67 MFLX40257-72 MFLX40257-73 MFLX40257-74 MFLX40257-80 MFLX40257-81 MFLX40257-82
Masterflex® 3/4" Sanitary Clamp High Flow Male Quick-Disconnect Fittings, Avantor®
Fittings and Connectors
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a High Flow Female fitting.
  • Aseptic connect and disconnect
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass through at lower line pressures in a smaller body. As the overall quick disconnect design has matured,More Product Information