Total-Guard Stem Casters, Polymer/Stainless Steel, Metro
Supplier: Metro
5PSTEB 5PSTE 5PSTEBX 5PSTEX
76545-628EA 315.26 CAD
76545-628 76545-630 76545-656 76545-658
Total-Guard Stem Casters, Polymer/Stainless Steel, Metro
Casters
Polymer/stainless steel total-guard stem casters are designed to provide excellent corrosion protection and moisture resistance for laboratory and clinical environments.
  • Excellent corrosion protection and moisture resistance
  • Non-marking thermoplastic elastomer (TPE) tread (70 shore) is quieter and more shock absorbent than a traditional polyurethane (TPU) tread
  • Total lock brake mechanism locks the wheel and swivel simultaneously
High-performance stem casters are great for laboratory and clinical storage applications. These casters are constructed ofMore Product Information