Saint Gobain® Tygon® A-60-F IB Pressure Transfer Tubing
Supplier: Saint Gobain Life Sciences
APW00029 APW00038 APW00019 MFLX95715-05
MFLX95715-01EA 883.22 CAD
MFLX95715-01 MFLX95715-02 MFLX95715-06 MFLX95715-05
Saint Gobain® Tygon® A-60-F IB Pressure Transfer Tubing
Tubing Transfer Tubing
Flexibility at extreme temperatures.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
Ideal for chemical transfer, pesticide/fertilizer delivery, water feed lines, food/dairy processing, and water purification lines. The tube features excellent resistance to acidic and alkali substances, ozone, UV light, and abrasive media. It is nonconductive, heat-sealable, non-aging, and nonoxidizing. In addition, the tube maintains flexibility even at extreme temperatures.
Temperature range is -60 to 275 °F (-51 to 135 °C).
Can be sterilized with autoclave.