CPC® Quick-Disconnect Fittings, 1⁄4-28 Flat-Bottom Port Insert
Supplier: VWR International
MFLX06368-71 PMCD48042812 (25PK)
MFLX06368-71CS 0 CAD
MFLX06368-71 MFLX06368-71BLK
CPC® Quick-Disconnect Fittings, 1⁄4-28 Flat-Bottom Port Insert
Fittings and Connectors
No more struggling—just press thumb latch.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-More Product Information