Reducer Fittings, Compression, Straight
Supplier: MIDWEST VALVE & FITTING INC
ISSD6RU5 ISSD4RU2 IBD8RU4 15-6-4-P-PG IBD5RU4 15-8-6-K-PG ISSD5RU4 IBD6RU4 IBD12RU8 15-10-6-K-PG ISSD6RU4 15-4-2-K-O IBD8RU6 15-5-4-K-PG 15-6-4-K-PG ISSD8RU4 15-6-4-N-PG
MFLX06381-83EA 86.43 CAD
MFLX06381-83 MFLX31412-25 MFLX31412-58 MFLX31107-30 MFLX06381-82 MFLX31412-56 MFLX06381-81 MFLX06381-80 MFLX31412-54 MFLX31406-23 MFLX31406-26 MFLX31406-27 MFLX31406-24 MFLX31406-25 MFLX06381-84 MFLX30803-60 MFLX31412-28
Reducer Fittings, Compression, Straight
Fittings and Connectors
Excellent choice to connect rigid tubing systems.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rates to 220 psi at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).