Union Fittings, Compression, Straight
Supplier: MIDWEST VALVE & FITTING INC
ISSD1U ISSD5U IBD4U IBD1U
MFLX06381-90EA 48.03 CAD
MFLX06381-90 MFLX06381-92 MFLX06381-93 MFLX31406-00 MFLX31406-50 MFLX31412-00 MFLX31412-50 MFLX06381-91 MFLX06381-94 MFLX06381-95 MFLX31406-01 MFLX31406-02 MFLX31406-03 MFLX31406-04 MFLX31406-51 MFLX31412-02 MFLX31412-51
Union Fittings, Compression, Straight
Fittings and Connectors
Get a secure seal at higher pressures.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rates to 220 psi at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).