Adapter Fittings, Compression to Male Threaded, Straight
Supplier: MIDWEST VALVE & FITTING INC
10-6-4-K-PG 10-10-6-K-PG 10-5-4-K-PG 10-12-8-K-PG 10-2-2-K-O 10-4-4-K-PG 10-12-12-K-PG 10-4-2-K-PG ISSD1MC1 10-8-8-K-PG 10-6-6-K-PG 10-10-8-K-PG 10-8-6-K-PG
MFLX06385-89EA 31.92 CAD
MFLX06385-89 MFLX06385-88 MFLX31406-33 MFLX06385-81 MFLX06385-80 MFLX06385-91 MFLX06385-83 MFLX06385-82 MFLX06385-85 MFLX06385-84 MFLX06385-87 MFLX06385-86 MFLX06385-90
Adapter Fittings, Compression to Male Threaded, Straight
Fittings and Connectors
Excellent choice to connect rigid tubing systems.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.