Fittings, Compression Nut Assembly with Ferrule
Supplier: MIDWEST VALVE & FITTING INC
MFLX06380-05 MFLX06380-61 MFLX06380-65 ISSD8FS+N ISSD6FS+N ISSD4FS+N ISSD2FS+N ISSD1FS+N IBD1FS+N IBD2FS+N IBD4FS+N IBD6FS+N MFLX06380-42 MFLX06380-60
MFLX06380-05EA 13.43 CAD
MFLX06380-05 MFLX06380-61 MFLX06380-65 MFLX31212-63 MFLX31212-64 MFLX31212-65 MFLX31212-66 MFLX31212-67 MFLX31212-68 MFLX31212-69 MFLX31212-70 MFLX31212-71 MFLX06380-42 MFLX06380-60
Fittings, Compression Nut Assembly with Ferrule
Fittings and Connectors
Can handle pressures up to 225 psi.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery