Adapter Fittings, Hose Barb to Female NPT Threaded, Straight
Supplier: MICROMOLD PRODUCTS, INC.
F-FH0102 F-FH0201 F-FH0202 F-FH0404 F-FH0203 F-FH0303 F-FH0403 F-FH0302 F-FH0304 F-FH0402 F-FH0606 F-FH0806 F-FH0808 F-FH0101
MFLX34030-89EA 165.9 CAD
MFLX34030-89 MFLX34030-90 MFLX34030-91 MFLX34030-98 MFLX34030-92 MFLX34030-94 MFLX34030-97 MFLX34030-93 MFLX34030-95 MFLX34030-96 MFLX34031-00 MFLX34031-01 MFLX34031-02 MFLX34030-88
Adapter Fittings, Hose Barb to Female NPT Threaded, Straight
Fittings and Connectors
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).