Bürkle Hand-Operated Pump Dispensers for Commercial Storage Drums
Supplier: Bürkle
5005-1200 5005-7000 5005-4000 5005-1500 5005-3000 5005-1000 5005-6000 0600-3003
MFLX06040-49EA 349.18 CAD
MFLX06040-49 MFLX06040-29 MFLX06040-51 MFLX06040-52 MFLX06040-53 MFLX06040-45 MFLX06040-46 MFLX06040-47
Bürkle Hand-Operated Pump Dispensers for Commercial Storage Drums
Pumps Drum/Transfer Pumps
Dispense liquids safely and quickly from most storage containers.
  • Telescopic immersion tube adapts itself to a variety of containers
  • Different materials and styles suit a wide range of applications
These hand-operated pumps feature a universal soft PVC bung consisting of a conical center and two conical sleeves allowing for an airtight seal between the pump and most liquid storage container openings. The telescopic immersion tube adapts itself to the container height on insertion, ensuring good removal of residual liquid. Various tube diameters are available to meet your individual liquid transfer needs. Choose from polypropylene (PP), PVDF, or 304 stainless steel (SS) wetted materials and dispensing by spout or hose.