LEVIFLOW® Single-Use Clamp-On Ultrasonic Flow Sensors
Supplier: LeviFlow
100-30390 100-30396
MFLX24500-19EA 4376.6 CAD
MFLX24500-19 MFLX24500-18
LEVIFLOW® Single-Use Clamp-On Ultrasonic Flow Sensors
Fluid Sensors
No moving parts—no particle generation—no contamination.
  • Starter kit and flow sensor required to build a complete system
  • Use on your existing silicone or C-Flex tubing
  • Obtain flow measurement without intrusion
  • Reusable flow sensors—choose from 4-, 20-, or 80-LPM models
These flow systems are ideal for sterile applications. Flow sensors fit over your existing silicone or C-Flex tubing lines and are easily reusable—remove sensor clamp, dispose of flexible tubing, and fix sensor over a new line.
These non-invasive flow measurement systems have no moving parts to contaminate or generate particulates, and are accurate to 3%.
Delivery information: Includes water tight connection box.