LEVIFLOW® Single-Use Precision Ultrasonic Flowmeters
Supplier: LeviFlow
100-30394 100-30375 100-30399 100-30379 100-30377 100-30400 100-30406 100-30397 100-30404 MFLX24500-15
MFLX24500-12EA 764.72 CAD
MFLX24500-12 MFLX24500-08 MFLX24500-09 MFLX24500-16 MFLX24500-10 MFLX24500-11 MFLX24500-13 MFLX24500-14 MFLX24500-17 MFLX24500-15
LEVIFLOW® Single-Use Precision Ultrasonic Flowmeters
Fluid Sensors
Obtain flow measurements of high-purity fluids.
  • Order the starter kit and a flow sensor to build a complete system
  • Choose from sterile or non-sterile flow sensors covering a range of 80 ml/min to 80 LPM
  • Avoid contamination— systems don't contain moving or obtrusive parts
  • Sensors feature integrated sanitary clamp fittings
  • Easily configure flow sensor parameters via software
These ultrasonic flowmeter systems are designed for use in applications were cleanliness is required. Readings are accurate to 1%—high precision for high-purity fluids. Each system requires a starter kit and clamp-on flow sensor; order separately. The starter kit includes components to get a system in place, just choose a flow sensor at the desired flow range and sterility.
Delivery information: Includes water tight connection box.