Almatec Air-driven Postive Displacement Pumps
Supplier: PSG CALIFORNIA LLC
E10TTT MFLX75980-16 MFLX76980-02 MFLX77980-01 MFLX77980-07
MFLX75980-00EA 2464.8 CAD
MFLX75980-00 MFLX75980-02 MFLX75980-04 MFLX75980-16 MFLX76980-02 MFLX77980-01 MFLX77980-07
Almatec Air-driven Postive Displacement Pumps
Pumps Air-Operated Pumps
German precision engineered AODD pumps to get your application up and flowing.
  • Metal-free air-operated double diaphragm pumps
The heart of the AODD is the air-valve and thoughtful German engineering went into the development of the precision control PERSWING P® air valve and its Air Distribution System (ADS). Resulting in a metal-free, pneumatically pilot-operated PERSWING P® (ADS) that ensures accurate reversal of the main piston and offers low noise levels, minimal maintenance and lube-free operation (due to two moving parts).
Almatec plastic pumpsMore Product Information