Sani-Tech® Ultra Platinum-Cured Silicone Lab Tubing
Supplier: ST. GOBAIN - TAUNTON
ULTRA-C-062-1 ULTRA-C-093-2 ULTRA-C-093-3H ULTRA-C-125-1H ULTRA-C-125-2H ULTRA-C-312-4F ULTRA-C-875-4 ULTRA-C-125-1F
MFLX96201-19EA 91.01 CAD
MFLX96201-19 MFLX96201-50 MFLX96201-55 MFLX96201-60 MFLX96201-63 MFLX96202-21 MFLX96202-97 MFLX96201-59
Sani-Tech® Ultra Platinum-Cured Silicone Lab Tubing
Tubing Transfer Tubing
Don't let extractables ruin your formulations.
  • Exceptionally low TOCs minimize fluid contamination
  • Ultra-pure silicone will not impart flavors or odors
  • Superior pumping life reduces downtime for tubing changes
Applications: Sterile fill and transfer, cell harvest, media processing, sanitary peristaltic pumping, production filtration and fermentation, bioreactor process lines, and pharmaceutical delivery.
Characteristics: Excellent flexibility, long pump life, non-yellowing, tasteless and odorless, nontoxic, translucent.
Certifications: Elastomer is FDA-compliant and meets USP Class VI, EP 3.1.9, and ISO 10993 biocompatibility criteria. Tubing is manufactured and packaged in a certified cleanroom.
Temperature range: -80 to 500 °F (-62 to 260 °C).
Sterilize: Autoclave, ethylene oxide, or gamma irradiation (up to 5.0 Mrad).