Harwil In-Line Liquid Level Switches
Supplier: VWR International
MFLX34001-55 MFLX34001-57 MFLX34001-59 MFLX34001-61 MFLX34001-63 MFLX34001-65
MFLX34001-55EA 432.92 CAD
MFLX34001-55 MFLX34001-57 MFLX34001-59 MFLX34001-61 MFLX34001-63 MFLX34001-65
Harwil In-Line Liquid Level Switches
Level Sensors
Eliminate nuisance switch tripping with a wide range of differentials.
  • Large differentials are immune to wave action and turbulence (L-21VCR models)
  • Single-point switch provides pump up/pump down level control
  • Withstands corrosive and particle-contaminated working fluids
This series of liquid level switches are easy to install, operate and maintain. Through-wall NPT process fittings and NPT electrical ports facilitate integration into existing systems. Operation checks and troubleshooting require only a multimeter.
Wetted materials are fiberglass-reinforced PPS and Hastelloy® C. Switch types and materials of construction resist corrosion and increase durability, making them suitable even for seawater and filtered sewage.