Catalyst Pump Tubing Links / Assemblies for FH15, FH30 and FH100M Pumps
Supplier: VWR International
MFLX97100-13 MFLX97100-16 MFLX97100-17 MFLX97100-18 MFLX97100-22 MFLX97100-23 MFLX97100-26 MFLX97100-42 MFLX97100-43 MFLX97100-46 MFLX97100-47 MFLX97100-63 MFLX97100-66 MFLX97100-67 MFLX97200-04 MFLX97200-06 MFLX97200-08 MFLX97200-23 MFLX97200-26 MFLX97200-27 MFLX97200-68 MFLX97100-27 MFLX97100-48 MFLX97200-62 MFLX97200-63 MFLX97200-64 MFLX97200-66 MFLX97200-67 MFLX97100-12
MFLX97100-13EA 330.22 CAD
MFLX97100-13 MFLX97100-16 MFLX97100-17 MFLX97100-18 MFLX97100-22 MFLX97100-23 MFLX97100-26 MFLX97100-42 MFLX97100-43 MFLX97100-46 MFLX97100-47 MFLX97100-63 MFLX97100-66 MFLX97100-67 MFLX97200-04 MFLX97200-06 MFLX97200-08 MFLX97200-23 MFLX97200-26 MFLX97200-27 MFLX97200-68 MFLX97100-27 MFLX97100-48 MFLX97200-62 MFLX97200-63 MFLX97200-64 MFLX97200-66 MFLX97200-67 MFLX97100-12
Catalyst Pump Tubing Links / Assemblies for FH15, FH30 and FH100M Pumps
Tubing Peristaltic Pump Tubing
Start pumping with little effort - preconfigured Links for quick and easy setup.
  • Four formulations offer wide range of chemical compatibility
  • Save time with already assembled links
  • Use with the Catalyst extenstion coils / tubing to complete your system
  • FH15 and FH30 utilize the stopped assemblies that have an ID of: 0.8, 1.6, 3.2, and 4.8 mm
  • FH100M (with the large cartridges) can use assemblies with an ID of: 0.8, 1.6, 3.2, 4.8, and 6.4 mm
  • FH100M (with the small cartridges) can use assemblies with an ID of: 0.16, 0.25, 0.89, 1.42, 2.06, and 2.79 mm