Noshok 500 Series Precision Needle Valves
Supplier: VWR International
MFLX06292-35 MFLX06292-36 MFLX06292-42 MFLX06292-43 MFLX06292-37 MFLX06292-39 MFLX06292-41 MFLX06292-44 MFLX06292-45
MFLX06292-35EA 94.33 CAD
MFLX06292-35 MFLX06292-36 MFLX06292-42 MFLX06292-43 MFLX06292-37 MFLX06292-39 MFLX06292-41 MFLX06292-44 MFLX06292-45
Noshok 500 Series Precision Needle Valves
Valves Needle Valves
Soft-seat design.
  • Soft seat design is 100% helium leak-tested to 1 x 10–4 ml/s at 200 psi
  • Designed with Delrin® acetal seat
  • Straight-through porting for bidirectional, high capacity flow and easy roddable cleaning
Series 500 needle valves are high quality instrument grade valves. They are the right choice when your application calls for positive shut-off or precise flow control. They work effectively with most gases and liquids.