Cadence Science Nickel-Plated Brass Luer Stopcocks
Supplier: CADENCE INC.
6019IND 6021IND 6023IND 6001IND 6010IND 6011IND 6014IND MFLX41507-15
MFLX41507-00EA 165.9 CAD
MFLX41507-00 MFLX41507-01 MFLX41507-02 MFLX41507-10 MFLX41507-11 MFLX41507-12 MFLX41507-13 MFLX41507-15
Cadence Science Nickel-Plated Brass Luer Stopcocks
Valves Ball Valves
Exceptional durability.
  • Easy to disassemble for cleaning
  • Stopcock handles turn 180°
  • Autoclavable
These Cadence Science luer stopcocks are constructed of nickel-plated brass for enhanced durability. These stopcocks offer worry-free cleaning with easy disassembly to save valuable time.