Masterflex® Smart Clamp® Stainless Steel Sanitary Clamps, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
SMARTCLAMP-300 SMARTCLAMP-200
MFLX61020-82EA 282.82 CAD
MFLX61020-82 MFLX31608-91
Masterflex® Smart Clamp® Stainless Steel Sanitary Clamps, Avantor®
Clamps Tubing and Sanitary Clamps
Torque control for consistent sealing of fittings.
  • Constructed of durable stainless steel
These sanitary clamps feature a unique design that allows fast clamping while reducing the repetitive strain injuries in hands and fingers that can occur with regular sanitary clamps; only a half turn is required. Clamps also have an adjustable torque for compression control and feature multiple settings for use with a variety of sanitary gaskets. Threadless design prevents soil and bacteria entrapment. Clamps also support ASME-BPE standards.