Masterflex® Color-Coded Luer Fittings, Polyolefin, Avantor®
Supplier: Avantor Fluid Handling
LMSLN-4 CVU-R 25PK LMSLN-2.5 CVU-R 25PK LMSLN-2.5 CVU-V 25PK LMSLN-4 CVU-L 25PK LMSLN-1.5 CVU-L 25PK LMSLN-1 CVU-L 25PK LMSLN-1.5 CVU-R 25PK LMSLN-1.5 CVU-B 25PK LMSLN-1.5 CVU-O 25PK LMSLN-2.5 CVU-Y 25PK LMSLN-2 CVU-Y 25PK LMSLN-1 CVU-R 25PK LMSLN-1 CVU-V 25PK LMSLN-2 CVU-L 25PK LMSLN-4 CVU-O 25PK LMSLN-2.5 CVU-B 25PK LMSLN-2.5 CVU-L 25PK LMSLN-1 CVU-Y 25PK LMSLN-3 CVU-R 25PK LMSLN-3 CVU-B 25PK LMSLN-1 CVU-O 25PK LMSLN-2 CVU-B 25PK LMSLN-1.5 CVU-V 25PK LMSLN-3 CVU-L 25PK LMSLN-3 CVU-V 25PK LMSLN-2 CVU-O 25PK LMSLN-2 CVU-V 25PK LMSLN-4 CVU-Y 25PK LMSLN-2 CVU-R 25PK LMSLN-3 CVU-Y 25PK LMSLN-1 CVU-B 25PK LMSLN-3 CVU-O 25PK LMSLN-4 CVU-B 25PK LMSLN-4 CVU-V 25PK LMSLN-1.5 CVU-Y 25PK
MFLX45520-42EA 42.5 CAD
MFLX45520-42 MFLX45520-21 MFLX45520-43 MFLX45520-41 MFLX45520-35 MFLX45520-13 MFLX45520-14 MFLX45520-36 MFLX45520-11 MFLX45520-33 MFLX45520-55 MFLX45520-12 MFLX45520-56 MFLX45520-34 MFLX45520-15 MFLX45520-16 MFLX45520-31 MFLX45520-53 MFLX45520-32 MFLX45520-51 MFLX45520-52 MFLX45520-02 MFLX45520-24 MFLX45520-46 MFLX45520-03 MFLX45520-25 MFLX45520-44 MFLX45520-22 MFLX45520-23 MFLX45520-45 MFLX45520-01 MFLX45520-06 MFLX45520-04 MFLX45520-26 MFLX45520-05
Masterflex® Color-Coded Luer Fittings, Polyolefin, Avantor®
Fittings and Connectors
Quick and easy assembly without tools.
  • Constructed of durable Polyolefin
Available in 6 colors for quick and easy identification: berry, rose, yellow, lime, violet, and orange. Made of polyolefin, a transparent polycarbonate alternative containing no bisphenol-A (BPA).
Superior chemical resistance compared to polycarbonate. Sterilize by autoclave, gamma irradiation, or ethylene oxide. Meets USP Class VI. Animal-derived ingredient-free. Packs of 25.