CPC® AseptiQuik® S Series Genderless Aseptic Connectors, Low Flow
Supplier: Colder Products
AQS33012HT AQS17002 AQS17004 AQS17006 AQS17004HT AQS17002HT AQS17006HT
MFLX20002-93EA 90.85 CAD
MFLX20002-93 MFLX20002-96 MFLX20202-48 MFLX20202-50 MFLX20202-52 MFLX20202-51 MFLX20202-49 MFLX20202-53
CPC® AseptiQuik® S Series Genderless Aseptic Connectors, Low Flow
Fittings and Connectors
Provide easy, secure, sterile connections even in nonsterile environments.
  • Genderless design simplifies system integration—no need to worry about female and male threads
  • Robust construction provides reliable performance—even without clamps
  • Quick, three-step connection process reduces risk of operator error
Manufactured from sturdy polycarbonate, these genderless connectors are ideal for single-use and sanitary applications. Use hose barb connectors with filters, tubing, and process containers to create systems for upstream or downstream processes.
Simply fit the connector half in your tubing line. The easy three-step FLIP-CLICK-PULL assembly procedure ensures a tight seal with no additional hardware required. The audible “click” confirms a securely connected assembly.
All fittings can be sterilized by gamma irradiation. Use high-temperature fittings to sterilize via autoclave.