Hand-Operated Water Transfer Pump, 6 strokes/gallon
Supplier: BECKSON INDUSTRIAL PRODUCTS
136PF-6
MFLX70607-14EA 126.87 CAD
MFLX70607-14
Hand-Operated Water Transfer Pump, 6 strokes/gallon
Pumps Drum/Transfer Pumps
Ideal for removing water in municipal and irrigation markets in addition to facilities maintenance.
  • Recommended for aqueous solutions
  • Withstand temperatures up to 120F (49C)
  • Self-priming, will not siphon
This hand-operated drum pump is ideal for pumping water and aqueous solutions. Constructed with a PVC body, hose and shaft, this pump offers 6 strokes/gallon.