Hydrogen (H2) Gas Mass Flow Controllers with Metal bodies
Supplier: AALBORG INSTRUMENTS & CONTROL
GFCS-010028 GFCS-010031 GFCS-010853 GFCS-010035 GFCS-010038 MFLX32660-56 MFLX32660-62 MFLX32660-64 MFLX32660-68 MFLX32660-71 MFLX32661-58 MFLX32661-60 MFLX32661-62 MFLX32661-64 MFLX32661-66 MFLX32661-52
MFLX32660-52EA 1674.8 CAD
MFLX32660-52 MFLX32660-58 MFLX32660-60 MFLX32660-66 MFLX32660-72 MFLX32660-56 MFLX32660-62 MFLX32660-64 MFLX32660-68 MFLX32660-71 MFLX32661-58 MFLX32661-60 MFLX32661-62 MFLX32661-64 MFLX32661-66 MFLX32661-52
Hydrogen (H2) Gas Mass Flow Controllers with Metal bodies
Flow Switches
Precisely control flow rates with built-in valve.
  • Proven design uses metal body components for added durability
  • Accumulated flow can be logged using an optional totalizer
  • Includes NIST-traceable certificate
These controllers feature an advanced U-shaped sensor that ensures accurate and repeatable results. Gas flow measurements are unaffected by moderate temperature and pressure variations at the inlet. The meter also allows a four-point calibration across the flow range to improve meter linearity.
The flow rate set point can be established by either aMore Product Information