Masterflex® Sani-Lock 316L Stainless Steel Quick-Connect Clamps, Avantor®
Supplier: VWR International
MFLX31607-70 MFLX31607-76 MFLX31607-84
MFLX31607-64EA 620.94 CAD
MFLX31607-64 MFLX31607-70 MFLX31607-76 MFLX31607-84 MFLX31607-60 MFLX31607-86
Masterflex® Sani-Lock 316L Stainless Steel Quick-Connect Clamps, Avantor®
Fittings and Connectors
No hinge clamp required – rapid one-hand connection.
  • Snap-in-place locking arms secure fitting, while safety pins prevent accidental release
  • Fully compatible with sanitary Tri-Clamp® fittings
  • 150 psi (10.3 bar) working line pressure
Converts any connection into a sanitary 'quick-connect' connection. Sani-Lock provides the convenience and efficiency of a quick-connect while maintaining the integrity of a sanitary connection. Use with Tuf-Flex® gaskets for guaranteed leak-proof alignment.(sold separately below) Automatic alignment of the Tuf-Flex gasket eliminates over and under compression of gaskets. The locking arms snap-in-place while creating a secure connection.