Gauge Guard Protector and Isolator Seals
Supplier: VWR International
40GGI-E-150 42GGP-PX-075 40GGP-PX-150 40GGI-E-200
MFLX30565-15EA 133.04 CAD
MFLX30565-15 MFLX30565-19 MFLX30565-01 MFLX30565-03 MFLX30565-21
Gauge Guard Protector and Isolator Seals
Seals Gaskets
Extend the life and accuracy of your expensive and fragile instruments.
  • Manufactured to meet USP Class VI certification
  • Animal derived ingredient free
Gauge Guard Protectors safeguard your expensive instruments from damaging process fluids and destructive routine maintenance such as autoclaving. Gauge guard is platinum-cured silicone combined with a 316 stainless steel perforated plate, and slips over the OD of your diaphragm seal. Model 30565-03 fits both 1 and 1 1/2" seals.
Gauge Guard Isolator Seals combine a hygenic seal with a membrane to isolate hot or caustic solutions from your instrument. Available in platinum-cured silicone or EPDM.