TriClamp Sanitary Strainer Assemblies, 316 Stainless Steel
Supplier: RUBBERFAB GASKET & MOLDING
RFILS-150-S-.125-316L RFILS-200-S-.250-316L
MFLX29590-02EA 1990.8 CAD
MFLX29590-02 MFLX29590-14 MFLX29590-00 MFLX29590-04 MFLX29590-10 MFLX29590-12 MFLX29590-52 MFLX29590-60 MFLX29590-64
TriClamp Sanitary Strainer Assemblies, 316 Stainless Steel
Filters Strainers
Protect products and expensive process equipment from coarse or fine particulates.
  • 316 SS fluid contact surfaces are polished to a smooth, blemish free finish
  • Standard clamp sizes allow easy installation or retrofit
Sanitary strainers are ideal for particulate removal or pre-filtration in the processing of foods, beverages, pharmaceuticals, cosmetics, and other critical compounds. Housings are available in in-line and side-inlet configurations.
Delivery information: Strainer assemblies include housing, perforated core, internal cap and spring, assembly clamp and gasket. Order wire mesh overscreens of the desired mesh size separately.