AW-Lake Sight Flow Transmitters
Supplier: AW-LAKE COMPANY
CCB-05-B-ES VCB-10-B-ES MFLX32825-04 MFLX32825-10 MFLX32825-12 MFLX32825-18
MFLX32825-00EA 976.44 CAD
MFLX32825-00 MFLX32825-20 MFLX32825-04 MFLX32825-10 MFLX32825-12 MFLX32825-18
AW-Lake Sight Flow Transmitters
Flow Indicators
Output signal for local and remote indication.
  • Easily view flow through plastic lens
These sight flow sensors are designed for low-cost applications where visual indication and sensing output signals are required—available with 4 to 20 mA, pulse, relay, or 0 to 5 VDC output. Durable polypropylene body is compatible with a variety of fluids. Sensors offer a flow measuring accuracy of ±2% full-scale, and are capable of handling pressures up to 150 psig and temperatures up to 150 °F (66 °C).