American Machine & Tool Straight 316 SS Standard Centrifugal Pumps
Supplier: American Machine & Tool
5483-98 5485-98 553B-98 5473-98 5497-98 5533-98 5539-98 5481-98 5531-98 5487-98 5535-98 5495-98 5471-98
MFLX70761-08EA 1124.96 CAD
MFLX70761-08 MFLX70761-18 MFLX70761-20 MFLX70761-30 MFLX70761-22 MFLX70761-10 MFLX70761-32 MFLX70761-24 MFLX70761-02 MFLX70761-12 MFLX70761-04 MFLX70761-26 MFLX70761-06
American Machine & Tool Straight 316 SS Standard Centrifugal Pumps
Pumps Centrifugal Pumps
Investment cast stainless steel (SS) pumps are built sturdier for heavy-duty, high performance applications requiring excellent chemical resistance.
  • Ideal for low-pressure OEM and chemical industry applications
  • Rotate head in 90° increments for easy installation of tubing
  • Available with totally enclosed fan cooled (TEFC) motors
  • Chemical resistant design
These stainless steel straight centrifugal pumps are designed for continuous duty high flow and high pressure applications including chemical processing, liquid transfer, cooling, pressure boosting, circulating and applications requiring contaminant free pumping.
Constructed of rugged deep drawn stainless steel for durability and corrosion resistance. Investment cast stainless steel closed impellers for enhanced performance and high efficiencies. All models include Viton® Type 21 mechanical seals and O-rings which offer a broad range of chemical compatibility. Cast iron motor adapter provides a positive and rigid mounting to motor.