PTFE Sanitary Clamp to Threaded Adapters
Supplier: MICROMOLD PRODUCTS, INC.
F-TCA1212 F-TCA1616 F-TCA0202 F-TCA0404
MFLX34031-09EA 598.82 CAD
MFLX34031-09 MFLX34031-10 MFLX34031-04 MFLX34031-06
PTFE Sanitary Clamp to Threaded Adapters
Fittings and Connectors
Create a secure connection in your sanitary fitting system.
  • Constructed of chemically inert PTFE
Chemically inert, PTFE offers outstanding resistance for any use, especially for critical high-purity applications. When subjected to higher pressure, PTFE as a material has a tendency to creep over time. As a result, these fittings work best at lower operating pressures (110 psi maximum operating pressure at ambient temperature for smaller sizes). All fittings are made from virgin PTFE material.