PTFE Reducing Coupler Fittings, Female NPT
Supplier: MICROMOLD PRODUCTS, INC.
F-RC0301 F-RC0402 F-RC0201 F-RC0302 F-RC0401 F-RC0602 F-RC0403
MFLX34030-36EA 191.18 CAD
MFLX34030-36 MFLX34030-35 MFLX34030-37 MFLX34030-32 MFLX34030-31 MFLX34030-34 MFLX34030-33
PTFE Reducing Coupler Fittings, Female NPT
Fittings and Connectors
Chemically-inert PTFE handles most aggressive chemicals.
  • Constructed of durable PTFE
Chemically inert, PTFE offers outstanding resistance for any use, especially for critical high-purity applications. When subjected to higher pressure, PTFE as a material has a tendency to creep over time. As a result, these fittings work best at lower operating pressures (110 psi maximum operating pressure at ambient temperature for smaller sizes). All fittings are made from virgin PTFE material.