Union Fittings, Female Threaded, Straight, PTFE
Supplier: MICROMOLD PRODUCTS, INC.
F-SC01 F-SC04 MFLX34030-27 MFLX34030-29 MFLX34030-30
MFLX34030-25EA 77.42 CAD
MFLX34030-25 MFLX34030-28 MFLX34030-27 MFLX34030-29 MFLX34030-30
Union Fittings, Female Threaded, Straight, PTFE
Fittings and Connectors
Chemically-inert PTFE handles most aggressive chemicals.
  • Constructed of durable PTFE
Chemically inert, PTFE offers outstanding resistance for any use, especially for critical high-purity applications. When subjected to higher pressure, PTFE as a material has a tendency to creep over time. As a result, these fittings work best at lower operating pressures (110 psi maximum operating pressure at ambient temperature for smaller sizes). All fittings are made from virgin PTFE material.