Union Fittings, Female NPT, 90° Elbow, PTFE
Supplier: MICROMOLD PRODUCTS, INC.
F-90E02 F-90E04 MFLX34030-07 MFLX34030-12
MFLX34030-08EA 99.7 CAD
MFLX34030-08 MFLX34030-10 MFLX34030-07 MFLX34030-12
Union Fittings, Female NPT, 90° Elbow, PTFE
Fittings and Connectors
Chemically-inert PTFE handles most aggressive chemicals.
  • Constructed of durable PTFE
Chemically inert, PTFE offers outstanding resistance for any use, especially for critical high-purity applications. When subjected to higher pressure, PTFE as a material has a tendency to creep over time. As a result, these fittings work best at lower operating pressures (110 psi maximum operating pressure at ambient temperature for smaller sizes). All fittings are made from virgin PTFE material.