Flowline EchoSonic II Ultrasonic Level Transmitters
Supplier: FLOWLINE
LU27-01 LU28-01 LU29-01
MFLX23002-02EA 2006.6 CAD
MFLX23002-02 MFLX23002-03 MFLX23002-04
Flowline EchoSonic II Ultrasonic Level Transmitters
Level Sensors
Non-contact level measurement.
  • Optimize sensor placement with unmatched beam width for applications with restricted space
  • Industry-leading miniature dead band maximizes the filling capacity of vessels
  • Offered in 3 measurement ranges up to 32.8 ft (10 m)
  • WebCal, an innovative PC user interface provides fast and accurate configuration
  • NEMA 4X rated polycarbonate enclosure rated with PVDF transducer
The EchoSonic II™ general purpose two-wire ultrasonic transmitter provides non-contact level measurement up to 32.8 ft (10 m), and is well suited forMore Product Information