CPC® Quick-Disconnect Fittings, Hose Barb Elbow Inserts
Supplier: VWR International
MFLX06361-27 MFLX06361-28 MFLX06361-39 MFLX06361-41 MFLX06361-63 MFLX06361-75 MFLX06361-76 MFLX06361-87 MFLX06361-98 MFLX06361-99 MFLX06362-00 MFLX06362-01 MFLX31307-67 MFLX31307-68 PMC2302 (25PK) PMC2304 (25PK) PMCD2302 (25PK) PMCD2304 (25PK) PLC23004 (25PK) PLCD23004 (25PK) PLCD23006 (25PK) PLC2300412 (25PK) PMCD230212 (25PK) PMCD230412 (25PK) PLCD2300412 (25PK) PLCD2300612 (25PK) PMC230412 PLC2300612 PLC23006
MFLX06361-27EA 14.85 CAD
MFLX06361-27 MFLX06361-28 MFLX06361-39 MFLX06361-41 MFLX06361-63 MFLX06361-64 MFLX06361-75 MFLX06361-76 MFLX06361-85 MFLX06361-87 MFLX06361-98 MFLX06361-99 MFLX06362-00 MFLX06362-01 MFLX31307-67 MFLX31307-68 MFLX06361-27BLK MFLX06361-28BLK MFLX06361-39BLK MFLX06361-41BLK MFLX06361-63BLK MFLX06361-75BLK MFLX06361-76BLK MFLX06361-87BLK MFLX06361-98BLK MFLX06361-99BLK MFLX06362-00BLK MFLX06362-01BLK MFLX31307-67BLK MFLX31307-68BLK MFLX06361-64BLK
CPC® Quick-Disconnect Fittings, Hose Barb Elbow Inserts
Fittings and Connectors
No more struggling—just press thumb latch.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-More Product Information