ATEX Special-Duty Air-Operated Double-Diaphragm Pumps
Supplier: WARREN RUPP
S05B2C1TCNS000 S05B2C2TCNS000
MFLX74440-70EA 2559.6 CAD
MFLX74440-70 MFLX74440-72
ATEX Special-Duty Air-Operated Double-Diaphragm Pumps
Pumps Air-Operated Pumps
Unique configurations to meet your demanding needs.
  • High flow rates — up to 106 GPM!
  • Air-powered motor eliminates sparking concerns associated with electrical pumps.
  • Bolted pump design allows easy access to pump head for maintenance and maintains seal after repeated servicing.
ATEX-Approved Nonmetallic Pumps provide 100% groundability with conductive materials of construction and ensuring no static electricity buildup for ignition-free operation. Pumps meet ATEX directives for potentially explosive atmospheres.