Stainless Steel Compression to NPT(M) Adapter Elbows
Supplier: MIDWEST VALVE & FITTING INC
ISSD4ME2 ISSD4ME4 ISSD6ME6 ISSD6ME4 ISSD2ME4 ISSD2ME2 ISSD6ME8
MFLX31406-77EA 36.97 CAD
MFLX31406-77 MFLX31406-78 MFLX31406-75 MFLX31406-76 MFLX31406-79 MFLX31406-80 MFLX31406-81
Stainless Steel Compression to NPT(M) Adapter Elbows
Fittings and Connectors
Create a secure connection with compression fittings.
  • Constructed of durable stainless steel (SS)
Very good chemical resistance. Pressure limitation determined by the tubing, not the fitting. Usable from -40 to 200ºF (-40 to 93ºC).