Stainless Steel Compression to NPT(M) Adapters
Supplier: MIDWEST VALVE & FITTING INC
ISSD2MC2 ISSD4MC4 ISSD6MC6 ISSD4MC8 ISSD4MC2 ISSD5MC6 ISSD6MC4 MFLX31406-38 MFLX31406-67 MFLX31406-68 MFLX31406-69 MFLX31406-70 MFLX31406-71 MFLX31406-72 MFLX31406-73 MFLX31406-74
MFLX31406-34EA 24.17 CAD
MFLX31406-34 MFLX31406-35 MFLX31406-36 MFLX31406-37 MFLX31406-64 MFLX31406-65 MFLX31406-66 MFLX31406-38 MFLX31406-67 MFLX31406-68 MFLX31406-69 MFLX31406-70 MFLX31406-71 MFLX31406-72 MFLX31406-73 MFLX31406-74
Stainless Steel Compression to NPT(M) Adapters
Fittings and Connectors
Excellent choice to connect rigid tubing.
  • Great chemical resistance
  • Easy to assemble the fitting to the tubing
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through the included sealed ferrule and using the nut to compress down to form a secure seal.
Very good chemical resistance. Pressure limitation determined by the tubing, not the fitting. Usable from -40 to 200ºF (-40 to 93ºC). Sterilize by either ethylene oxide, autoclave, or gamma irradiation.