Single-Phase Input Variable-Speed Controllers
Supplier: ADDISON ELECTRIC INC
ESV251N01SXB ESV371N01SXB ESV751N01SXB
MFLX70101-01EA 744.18 CAD
MFLX70101-01 MFLX70101-11 MFLX70101-21
Single-Phase Input Variable-Speed Controllers
Pumps Motors
Operate a 230-240 VAC motor with only a 115 VAC power supply.
  • Use with a variety of motors
  • An excellent option for most¡ 1-hp applications
  • On-board trimmer pot allows easy calibration and setup
These controllers have the ability to double their output voltage in situations where 230-240 VAC is needed but only a 115 VAC power supply is available. To protect your motor and equipment, a torque foldback feature allows a 200% torque maximum for a short period of time to overcome intermittent peak loads. Torque levels are then reduced back to a preset safe level. NEMA 4X controllers are available; call our Application Specialists to order.