PendoTECH® Single-Use Pressure Monitoring and Transmitting Systems
Supplier: PENDOTECH
PMAT2 PMAT2-BAR PMAT3 PMAT3-BAR PMAT4A PMAT4A-BAR PMAT4R PMAT3P PMAT3P-BAR PMAT-S PMAT-2P MFLX19407-00 MFLX19407-02 PMAT4R-BAR
MFLX19406-02EA 8705.8 CAD
MFLX19406-02 MFLX19406-03 MFLX19406-04 MFLX19406-05 MFLX19406-06 MFLX19406-07 MFLX19406-08 MFLX19406-10 MFLX19406-11 MFLX19406-40 MFLX19406-41 MFLX19407-00 MFLX19407-02 MFLX19406-09
PendoTECH® Single-Use Pressure Monitoring and Transmitting Systems
Fluid Sensors
Single-use solution to monitor process pressure.
  • Ideal for applications such as tangential flow filtration, cross flow filtration, depth filtration, bioreactor monitoring, and chromatography
  • Data output capability to a PC or control system
  • User-configurable Min/Max alarm set points
The PressureMAT combines a pressure monitor, data logger, alarm, and transmitter in one simple control box. Designed for use with single-use, in-line pressure sensors. Each channel takes pressure readings once per second. Process pressure is displayed on the easy-to-read LCD. System is easily calibrated to zero through a single button. The PressureMAT may also transmit output via 4-20 mA signal or can provide RS-232 data transfer for use in common PC applications, such as the PressureMAT Data Acquisition Software or Excel®, through WinWedge or Hyper Terminal software.