Hedland Flow Meters for Air and Other Compressed Gases

Supplier: BADGER METER INC
H271A-005-EG H271A-010 H671A-050 H671A-100 MFLX32206-37 MFLX32510-56 MFLX32510-59 MFLX32510-60 MFLX32510-63 MFLX32510-68 MFLX32510-76 MFLX32510-82 MFLX32510-87 MFLX32510-92 MFLX32511-00 MFLX32511-04 MFLX32511-24 MFLX32511-27 MFLX32511-31 MFLX32511-48
MFLX32510-57EA 731.54 CAD
MFLX32510-57 MFLX32510-58 MFLX32510-78 MFLX32510-80 MFLX32206-37 MFLX32510-56 MFLX32510-59 MFLX32510-60 MFLX32510-63 MFLX32510-68 MFLX32510-76 MFLX32510-82 MFLX32510-87 MFLX32510-92 MFLX32511-00 MFLX32511-04 MFLX32511-24 MFLX32511-27 MFLX32511-31 MFLX32511-48
Hedland Flow Meters for Air and Other Compressed Gases
Flow Meters
Direct read 360° rotatable scale.

  • Easy-to-read linear scale

FEATURES
Direct readings
Install in any position
360° Rotatable Guard/Scale
No Flow Straighteners or Special Piping Required
Insensitive to Shock and Vibration
Good Viscosity Stability
Temperature up to 240 °F
Accuracy +/-2% Full Scale
Repeatability +/-1%
Special Scales Available
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR