GF Signet® Paddle-Wheel Flow System Installation Fittings
Supplier: GF Signet
CS4W040 CS4W080 CS4W060 CS4T010 MPV8T010F P526-2080
MFLX32511-74EA 706.26 CAD
MFLX32511-74 MFLX32511-95 MFLX32512-26 MFLX32511-80 MFLX32511-83 MFLX32511-82
GF Signet® Paddle-Wheel Flow System Installation Fittings
Flow Meters
Build a complete paddle-wheel flow sensor system customizable to your application.
  • Modular system for a customizable approach to flow measurement
  • Ideal for easy installation
The GF Signet paddle-wheel flow system is comprised of three components. Select the flow sensor, installation fitting, and flow meter that suit your application needs for a customizable approach to flow measurement.
Step One: Choose a Flow Sensor
Step Two: Choose an Installation Fitting
Step Three: Choose a Flow Meter
Flow sensor, installation fitting, and flow meter sold separately.