John Guest acetal push-to-connect union T-connectors
Supplier: VWR International
PI0208S-US PI0212S-US PI0216S PM0204S PM0208S PP0208W-US PP0212W-US
MFLX34006-11CS 0 CAD
MFLX34006-11 MFLX34006-12 MFLX34006-13 MFLX34006-53 MFLX34006-54 MFLX34007-46 MFLX34007-47 MFLX34006-30
John Guest acetal push-to-connect union T-connectors
Fittings and Connectors
No need to buy two fitting halves.
  • Meet NSF and FDA compliance for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth— tubing slides right out.
Fittings work with semirigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing. To connect flexible tubing such as Tygon® or silicone, order soft tube supports.